Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Isoproterenol Sulfate Dihydrate 97%, Thermo Scientific™

Catalog No. AC146191000
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC146191000 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC146191000 Supplier Thermo Scientific Chemicals Supplier No. 146191000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6700-39-6
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H19NO7S
Linear Formula 3, 4-(HO)2C6H3CH(OH)CH2NHCH(CH3)2.H2SO4
Quantity 100g
MDL Number MFCD07371352
Merck Index 15, 5263
Synonym isoproterenol sulfate, medihaler iso, luf-iso, isoprenaline sulfate, isoprenaline sulfate dihydrate, dl-isoproterenol sulfate dihydrate, isoprenaline sulfate jan, isoproterenol sulfate usp, 1,2-benzenediol, 4-1-hydroxy-2-methylethyl amino ethyl-, sulfate 2:1 salt , dihydrate
InChI Key YTOSPBSEWLATJU-UHFFFAOYSA-N
SMILES CC(C)NCC(C1=CC(=C(C=C1)O)O)O.OS(=O)(=O)O
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid
Molecular Weight (g/mol) 309.333
PubChem CID 9796663
Formula Weight 556.64
Percent Purity ≥96.0% (on dry substance)
Chemical Name or Material DL-Isoproterenol sulfate dihydrate, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.