Learn More
Medchemexpress LLC DL-lactic acid | 50-21-5 | MFCD00004520 | 91.1% | 90.08 | C3H6O3 | 10 MG

Supplier: Medchemexpress LLC HYB2227R10MG
DL-lactic acid analytical standard supplied as a 10 mg reference material intended for research and analytical applications. It is used as a reference material for method validation, calibration, and metabolite analysis. For research use only; not for human or clinical use.
- Analytical standard for reference and calibration.
- Assay approximately 91.1%.
- Molecular formula C3H6O3, molecular weight 90.08.
- Supplied in small pack sizes suitable for analytical workflows.
- Not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.