Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL - Lactic Acid Lithium Salt, MP Biomedicals™

Catalog No. 0210082410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-100-82410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-100-82410 Supplier MP Biomedicals, Inc Supplier No. 0210082410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL - Lactic Acid Lithium Salt is an α hydroxyl carboxylic acid produced from pyruvate by the enzyme lactate dehydrogenase (LDH).

  • Lactic acid is an α hydroxyl carboxylic acid produced from pyruvate by the enzyme lactate dehydrogenase (LDH) solution.
  • It is a fermentation acid from milk or carbohydrates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-55-0
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
InChI Key GKQWYZBANWAFMQ-UHFFFAOYNA-M
IUPAC Name lithium(1+) 2-hydroxypropanoate
SMILES [Li+].CC(O)C([O-])=O

Specifications

Quantity 10 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.