Learn More
Medchemexpress LLC DL-Lactic acid (Standard) | 50-21-5 | 91.1% | 50 MG
SDP
Catalog No. 5000410109
Encompass_Preferred
DL-Lactic acid (Standard) is an analytical standard for Lactic acid (HY-B2227), intended for research and analytical applications. It acts as a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator, inducing lysine residues lactylation. As a glycolysis end-product, it links glycolysis with oxidative phosphorylation. This oncometabolite also plays an immune protective role in anti-tumor immunity and possesses antimicrobial activity, suitable for use as a food preservative.
  • Analytical standard for research and analytical applications.
  • Activates hydroxycarboxylic acid receptor 1 (HCAR1).
  • Induces lysine residues lactylation.
  • Glycolysis end-product, linking glycolysis and oxidative phosphorylation.
  • Immune protective role in anti-tumor immunity.
  • Antimicrobial activity; suitable as a food preservative.
  • Used in HPLC, GC, and MS for research.
Catalog No. 50-004-10109 Supplier Medchemexpress LLC Supplier No. HYB2227R50MG
DL-Lactic acid (Standard) is an analytical standard for Lactic acid (HY-B2227), intended for research and analytical applications. It acts as a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator, inducing lysine residues lactylation. As a glycolysis end-product, it links glycolysis with oxidative phosphorylation. This oncometabolite also plays an immune protective role in anti-tumor immunity and possesses antimicrobial activity, suitable for use as a food preservative.
  • Analytical standard for research and analytical applications.
  • Activates hydroxycarboxylic acid receptor 1 (HCAR1).
  • Induces lysine residues lactylation.
  • Glycolysis end-product, linking glycolysis and oxidative phosphorylation.
  • Immune protective role in anti-tumor immunity.
  • Antimicrobial activity; suitable as a food preservative.
  • Used in HPLC, GC, and MS for research.