Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-Lactonitrile

Catalog No. 6983025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-830-25ML 25 mL
69-830-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-830-25ML Supplier Sigma Aldrich Supplier No. 6983025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DL-Lactonitrile

Specifications

CAS 78-97-7
Density 0.991 g/mL (at 20°C (literature))
Boiling Point 90°C (17 mmHg)
Molecular Formula C3H5NO
Refractive Index n20/D 1.404
Linear Formula CH3CH(OH)CN
MDL Number MFCD00004507
Quantity 25 mL
Synonym 2-Hydroxypropionitrile; Acetaldehyde cyanohydrin
Molecular Weight (g/mol) 71.08
Percent Purity ≥97.0% (T)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.