Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dl-laudanosine | 1699-51-0 | 99.5% | 10 MG
SDP

Catalog No. 5000231496
Encompass_Preferred

Dl-laudanosine is a research-grade metabolite standard of the neuromuscular blocking agents atracurium and cisatracurium. It crosses the blood-brain barrier and is associated with central excitatory effects; intended for research and analytical use only.

  • Analytical standard for HPLC, GC, and MS.
  • Molecular formula C21H27NO4; molecular weight 357.44 g/mol.
  • Approximately 99.5% purity.
  • Available in small pack sizes, including 10 mg.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • SDS and certificate of analysis available.

Catalog No. 50-002-31496 Supplier Medchemexpress LLC Supplier No. HY12248910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dl-laudanosine is a research-grade metabolite standard of the neuromuscular blocking agents atracurium and cisatracurium. It crosses the blood-brain barrier and is associated with central excitatory effects; intended for research and analytical use only.

  • Analytical standard for HPLC, GC, and MS.
  • Molecular formula C21H27NO4; molecular weight 357.44 g/mol.
  • Approximately 99.5% purity.
  • Available in small pack sizes, including 10 mg.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • SDS and certificate of analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.