Learn More
Medchemexpress LLC DL-Leucyl-DL-phenylalanine | 56217-82-4 | 99.7% | 278.35 | 100 MG
SDP
Catalog No. 5000349417
Encompass_Preferred
DL-Leucyl-DL-phenylalanine is a dipeptide that can be utilized as a substrate to detect two regions of dipeptidase staining on a gel in Drosophila simulans and Drosophila melanogaster.
  • Dipeptide
  • Substrate for dipeptidase staining detection
  • Suitable for Drosophila simulans and Drosophila melanogaster research
Catalog No. 50-003-49417 Supplier Medchemexpress LLC Supplier No. HY139440100MG
DL-Leucyl-DL-phenylalanine is a dipeptide that can be utilized as a substrate to detect two regions of dipeptidase staining on a gel in Drosophila simulans and Drosophila melanogaster.
  • Dipeptide
  • Substrate for dipeptidase staining detection
  • Suitable for Drosophila simulans and Drosophila melanogaster research