Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Lysinamide Dihydrochloride, MP Biomedicals™

Catalog No. ICN15528791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15528791 1 g
ICN15528780 100 mg
ICN15528783 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15528791 Supplier MP Biomedicals, Inc Supplier No. 0215528791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 51127-08-3 | C6H17Cl2N3O | 218.12 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51127-08-3
Molecular Formula C6H17Cl2N3O
Molecular Weight (g/mol) 218.12
MDL Number MFCD00058287
InChI Key AIYVJLPYZQDCKV-XRIGFGBMSA-N
Synonym s-2,6-diaminohexanamide dihydrochloride, h-lys-nh2 2hcl, l-lysinamide dihydrochloride, h-lys-nh 2 hcl, lysine amide dihydrochloride, c6h15n3o.2hcl, h-lys-nh2 . 2 hcl
PubChem CID 20760365
IUPAC Name (2S)-2,6-diaminohexanamide dihydrochloride
SMILES Cl.Cl.NCCCC[C@H](N)C(N)=O

Specifications

Quantity 1 g
Formula Weight 218.1
Physical Form Solid
Chemical Name or Material DL-Lysinamide Dihydrochloride
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.