Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-Lysine | 70-54-2 | 99.7% | 25 G
SDP

Catalog No. 5000414097
Encompass_Preferred

DL-Lysine is a racemic mixture of D-Lysine and L-Lysine, both α-amino acids essential for protein biosynthesis. D-Lysine, an enantiomer of L-Lysine, is produced through successive chemical racemization and microbial asymmetric degradation. It is present in all living organisms, from bacteria to humans, and is considered a potentially toxic compound. This product is intended for research use only by experienced personnel in appropriately equipped facilities and is not validated for medical applications.

  • Racemic mixture of D-Lysine and L-Lysine
  • Utilized in protein biosynthesis
  • D-Lysine produced from L-Lysine by chemical racemization and microbial asymmetric degradation
  • Potentially toxic compound
  • Soluble in water at 100 mg/mL
  • Stable under recommended storage conditions
  • Target: Endogenous Metabolite
  • Pathway: Metabolic Enzyme/Protease

Catalog No. 50-004-14097 Supplier Medchemexpress LLC Supplier No. 25GHYB2236
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Lysine is a racemic mixture of D-Lysine and L-Lysine, both α-amino acids essential for protein biosynthesis. D-Lysine, an enantiomer of L-Lysine, is produced through successive chemical racemization and microbial asymmetric degradation. It is present in all living organisms, from bacteria to humans, and is considered a potentially toxic compound. This product is intended for research use only by experienced personnel in appropriately equipped facilities and is not validated for medical applications.

  • Racemic mixture of D-Lysine and L-Lysine
  • Utilized in protein biosynthesis
  • D-Lysine produced from L-Lysine by chemical racemization and microbial asymmetric degradation
  • Potentially toxic compound
  • Soluble in water at 100 mg/mL
  • Stable under recommended storage conditions
  • Target: Endogenous Metabolite
  • Pathway: Metabolic Enzyme/Protease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.