Learn More
Medchemexpress LLC Dl-lysine-d4 (dihydrochloride) | 284664-88-6 | 98.0% | 223.13 | C6H12D4Cl2N2O2 | 10 MG
SDP
Catalog No. 5000297308
Encompass_Preferred
DL-lysine-d4 dihydrochloride is a deuterium-labeled, racemic lysine salt supplied as the dihydrochloride for use as a tracer or internal standard in quantitative analyses such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled DL-lysine dihydrochloride (labels at positions 4,4,5,5)
  • Intended as a tracer and internal standard for quantitative analysis
  • Purity 98.0%
  • White to off-white solid; soluble in water (100 mg/mL) and DMSO (1 mg/mL)
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years)
Catalog No. 50-002-97308 Supplier Medchemexpress LLC Supplier No. HYB2236S10MG
DL-lysine-d4 dihydrochloride is a deuterium-labeled, racemic lysine salt supplied as the dihydrochloride for use as a tracer or internal standard in quantitative analyses such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled DL-lysine dihydrochloride (labels at positions 4,4,5,5)
  • Intended as a tracer and internal standard for quantitative analysis
  • Purity 98.0%
  • White to off-white solid; soluble in water (100 mg/mL) and DMSO (1 mg/mL)
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years)