Learn More
Cayman Chemical DL-MalIc AcId 50g
SDP
Catalog No. 502950937
Encompass_Preferred
The racemate of L-malic acid; reduces IFN-γ and TNF-α-induced CCL2 and ICAM expression in HaCaT human keratinocytes at 1 mM; inhibits epidermis and dermis thickening, as well as mast cell and eosinophil dermal infiltration in a mouse model of DNBC-induced atopic dermatitis at 10 mM
Catalog No. 50-295-0937 Supplier Cayman Chemical Supplier No. 3331050 G
The racemate of L-malic acid; reduces IFN-γ and TNF-α-induced CCL2 and ICAM expression in HaCaT human keratinocytes at 1 mM; inhibits epidermis and dermis thickening, as well as mast cell and eosinophil dermal infiltration in a mouse model of DNBC-induced atopic dermatitis at 10 mM