Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Malic Acid, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Catalog No. 02002755 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-755 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-755 Supplier Avantor Supplier No. P49407

missing translation for 'validMainframeMsg'

High quality biochemicals for research use

Chemical Identifiers

CAS 617-48-1
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
InChI Key BJEPYKJPYRNKOW-UHFFFAOYSA-N
Synonym malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
PubChem CID 525
ChEBI CHEBI:6650
IUPAC Name 2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Formula Weight 134.09
Grade BAKER ANALYZED™ Biochemical Reagent
Quantity 500 g
Packaging Poly Bottle
Specific Gravity 1L = 1.60kg
Chemical Name or Material DL-Malic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.