Learn More
Medchemexpress LLC DL-Mannitol | 87-78-5 | 10 G
SDP
Catalog No. 5000418783
Encompass_Preferred
DL-Mannitol is derived from combining D-mannitol with L-mannitol, obtained by the reduction of L-mannono-1, Clactone. It presents as a solid, stable under recommended storage conditions, and is primarily intended for laboratory research use.
  • White to light yellow solid.
  • Stable under recommended storage conditions.
  • Powder storage: -20°C for 3 years or 4°C for 2 years.
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month.
  • Soluble in H2O at 62.5 mg/mL (343.09 mM) with ultrasonic and warming.
  • Soluble in PBS at 50 mg/mL (274.47 mM) requiring ultrasonic for a clear solution.
Catalog No. 50-004-18783 Supplier Medchemexpress LLC Supplier No. HYN661810G
Add to Cart
Add to Cart
DL-Mannitol is derived from combining D-mannitol with L-mannitol, obtained by the reduction of L-mannono-1, Clactone. It presents as a solid, stable under recommended storage conditions, and is primarily intended for laboratory research use.
  • White to light yellow solid.
  • Stable under recommended storage conditions.
  • Powder storage: -20°C for 3 years or 4°C for 2 years.
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month.
  • Soluble in H2O at 62.5 mg/mL (343.09 mM) with ultrasonic and warming.
  • Soluble in PBS at 50 mg/mL (274.47 mM) requiring ultrasonic for a clear solution.