Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-Mannitol | 87-78-5 | 25 G
SDP

Catalog No. 5000422010
Encompass_Preferred

DL-Mannitol is a chemical product primarily intended for research use only and not for sale to patients. It is obtained by combining D-mannitol with a sample of L-mannitol, which is acquired through the reduction of L-mannono-1, C-lactone. It is identified as suitable for laboratory chemicals and the manufacture of various substances.

  • Assay of 99.6%
  • Molecular weight of 182.17
  • Chemical formula C6H14O6
  • Appearance as a white to light yellow solid
  • Classified as a saccharide
  • Derived from plants other families
  • For research use only
  • Suitable for laboratory and manufacturing applications

Catalog No. 50-004-22010 Supplier Medchemexpress LLC Supplier No. 25GHYN6618
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Mannitol is a chemical product primarily intended for research use only and not for sale to patients. It is obtained by combining D-mannitol with a sample of L-mannitol, which is acquired through the reduction of L-mannono-1, C-lactone. It is identified as suitable for laboratory chemicals and the manufacture of various substances.

  • Assay of 99.6%
  • Molecular weight of 182.17
  • Chemical formula C6H14O6
  • Appearance as a white to light yellow solid
  • Classified as a saccharide
  • Derived from plants other families
  • For research use only
  • Suitable for laboratory and manufacturing applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.