Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-Mannitol | 87-78-5 | 50 G
SDP

Catalog No. 5000418446
Encompass_Preferred

DL-Mannitol is a chemical compound formed by combining D-mannitol with L-mannitol, obtained through the reduction of L-mannono-1, Clactone. This product is a solid, typically appearing white to light yellow, with a molecular weight of 182.17 and a chemical formula of C6H14O6. It is intended for research use only.

  • Assay purity of 99.6%
  • Solid form, white to light yellow in color
  • Molecular formula C6H14O6
  • Water solubility: 62.5 mg/mL for in vitro use
  • PBS solubility: 50 mg/mL for in vivo use
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-18446 Supplier Medchemexpress LLC Supplier No. HYN661850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Mannitol is a chemical compound formed by combining D-mannitol with L-mannitol, obtained through the reduction of L-mannono-1, Clactone. This product is a solid, typically appearing white to light yellow, with a molecular weight of 182.17 and a chemical formula of C6H14O6. It is intended for research use only.

  • Assay purity of 99.6%
  • Solid form, white to light yellow in color
  • Molecular formula C6H14O6
  • Water solubility: 62.5 mg/mL for in vitro use
  • PBS solubility: 50 mg/mL for in vivo use
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.