Learn More
Apexbio Technology LLC DL-MENTHOL-20MG
SDP
Catalog No. 5000567494
Encompass_Preferred
DL-Menthol (CAS No 89-78-1) is a compound commonly used in biomedical research within the category of terpenes originating from the mint family It acts by interacting with transient receptor potential (TRP) channels particularly TRPM8 which leads to its well-known cooling sensation This involvement in TRP channel modulation makes it a subject of interest in studies related to pain inflammation and sensory perception In vitro it exhibits activity in the nanomolar to low micromolar range making it suitable for precise biochemical assays DL-Menthol is frequently employed in a variety of applications including cell models animal studies and drug screening processes Its experimental concentrations typically depend on the specific research design employed The compound s effects on cellular pathways and its therapeutic potential continue to be areas of active investigation in the context of neurology and pharmacology
Catalog No. 50-005-67494 Supplier Apexbio Technology LLC Supplier No. N187520
DL-Menthol (CAS No 89-78-1) is a compound commonly used in biomedical research within the category of terpenes originating from the mint family It acts by interacting with transient receptor potential (TRP) channels particularly TRPM8 which leads to its well-known cooling sensation This involvement in TRP channel modulation makes it a subject of interest in studies related to pain inflammation and sensory perception In vitro it exhibits activity in the nanomolar to low micromolar range making it suitable for precise biochemical assays DL-Menthol is frequently employed in a variety of applications including cell models animal studies and drug screening processes Its experimental concentrations typically depend on the specific research design employed The compound s effects on cellular pathways and its therapeutic potential continue to be areas of active investigation in the context of neurology and pharmacology