Learn More
Medchemexpress LLC DL-menthol | 89-78-1 | MFCD00001484 | 1mL
SDP
Catalog No. 5000266140
Encompass_Preferred
DL-Menthol (Racementhol) is an orally active GABAaR positive allosteric modulator and UDP-glucuronosyltransferase (UGT) inhibitor that can cross the blood-brain barrier DL-Menthol binds to GABAAR and exhibits an allosteric activation effect enhancing GABA-mediated chloride influx and inhibiting neuronal excitability DL-Menthol can induce surgical anesthesia in fish and inhibit the metabolic detoxification of tobacco carcinogens by human liver and intestinal UGT enzymes resulting in reduced NNAL-N-Gluc production[1][2]
Catalog No. 50-002-66140 Supplier Medchemexpress LLC Supplier No. HYY168310MM/1ML
DL-Menthol (Racementhol) is an orally active GABAaR positive allosteric modulator and UDP-glucuronosyltransferase (UGT) inhibitor that can cross the blood-brain barrier DL-Menthol binds to GABAAR and exhibits an allosteric activation effect enhancing GABA-mediated chloride influx and inhibiting neuronal excitability DL-Menthol can induce surgical anesthesia in fish and inhibit the metabolic detoxification of tobacco carcinogens by human liver and intestinal UGT enzymes resulting in reduced NNAL-N-Gluc production[1][2]