Learn More
Apexbio Technology LLC DL-METANEPHRINE-D3-HCL-1MG
SDP
Catalog No. 5000570824
Encompass_Preferred
DL-Metanephrine-d3 HCl (CAS No 1085333-94-3) is a deuterium-labeled compound used extensively in biomedical research This compound is instrumental in studying various biochemical pathways particularly those involving catecholamine metabolism As a stable isotope-labeled compound it aids in the precise quantification of metanephrines in biological samples The mechanism of action primarily involves mimicking endogenous compounds facilitating the understanding of their metabolic processes It exhibits in vitro activity typically in the nanomolar to low micromolar range Common applications include its use in cell models and animal studies especially in the context of drug screening and pharmacokinetic studies Experimental concentrations typically depend on the specific research design allowing researchers to customize conditions to their experimental requirements This versatility makes it a valuable tool in the development of therapeutic interventions and metabolic research
Catalog No. 50-005-70824 Supplier Apexbio Technology LLC Supplier No. SL10041
DL-Metanephrine-d3 HCl (CAS No 1085333-94-3) is a deuterium-labeled compound used extensively in biomedical research This compound is instrumental in studying various biochemical pathways particularly those involving catecholamine metabolism As a stable isotope-labeled compound it aids in the precise quantification of metanephrines in biological samples The mechanism of action primarily involves mimicking endogenous compounds facilitating the understanding of their metabolic processes It exhibits in vitro activity typically in the nanomolar to low micromolar range Common applications include its use in cell models and animal studies especially in the context of drug screening and pharmacokinetic studies Experimental concentrations typically depend on the specific research design allowing researchers to customize conditions to their experimental requirements This versatility makes it a valuable tool in the development of therapeutic interventions and metabolic research