Learn More
AdipoGen DL-Methionine
SDP
Catalog No. 502561074
Encompass_Preferred
Chemical. CAS 59-51-8. Formula C5H11NO2S. MW 149.21. Synthetic. Essential amino acid involved in various biochemical processes in humans and mammals. Important in angiogenesis, the growth of new blood vessels and supplementation may be beneficial in diseases such as Parkinsons, schizophrenia, asthma or allergies, as well as in radiation, copper poisoning, alcoholism or depressions. Through transmethylation it can be converted to S-adenosylmethionine, a primary methyl donor that methylates compounds to form such products as creatine and phosphatidylcholine. Methionine can be converted to cysteine, a precursor of glutathione and taurine. These molecules may regulate intestinal epithelial oxidative status, and may contribute to intestinal mucosal integrity and gut function. Overconsumption of methionine has been associated to cancer growth. Restriction of methionine can increase lifespans and inhibit cancer growth. Used in livestock nutrition.
Catalog No. 50-256-1074 Supplier AdipoGen Supplier No. CDXD0444G100
Chemical. CAS 59-51-8. Formula C5H11NO2S. MW 149.21. Synthetic. Essential amino acid involved in various biochemical processes in humans and mammals. Important in angiogenesis, the growth of new blood vessels and supplementation may be beneficial in diseases such as Parkinsons, schizophrenia, asthma or allergies, as well as in radiation, copper poisoning, alcoholism or depressions. Through transmethylation it can be converted to S-adenosylmethionine, a primary methyl donor that methylates compounds to form such products as creatine and phosphatidylcholine. Methionine can be converted to cysteine, a precursor of glutathione and taurine. These molecules may regulate intestinal epithelial oxidative status, and may contribute to intestinal mucosal integrity and gut function. Overconsumption of methionine has been associated to cancer growth. Restriction of methionine can increase lifespans and inhibit cancer growth. Used in livestock nutrition.