Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dl-methionine methylsulfonium (chloride) | 3493-12-7 | 1 KG
SDP

Catalog No. 5000420179
Encompass_Preferred

DL-Methionine methylsulfonium chloride is a naturally occurring methionine derivative used as a laboratory chemical and for the manufacture of substances. It protects gastric mucosal from ethanol-induced damage. This product is intended for research use only by experienced personnel.

  • Naturally occurring methionine derivative
  • Protects gastric mucosal from ethanol-induced damage
  • Solid appearance, white to off-white color
  • Purity of 98.0%
  • Molecular weight of 199.70
  • Chemical formula: C6H14ClNO2S
  • Melting/freezing point: 137-138°C
  • Soluble in water and PBS
  • Stable under recommended storage conditions
  • For research use only

Catalog No. 50-004-20179 Supplier Medchemexpress LLC Supplier No. HYN66551KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Methionine methylsulfonium chloride is a naturally occurring methionine derivative used as a laboratory chemical and for the manufacture of substances. It protects gastric mucosal from ethanol-induced damage. This product is intended for research use only by experienced personnel.

  • Naturally occurring methionine derivative
  • Protects gastric mucosal from ethanol-induced damage
  • Solid appearance, white to off-white color
  • Purity of 98.0%
  • Molecular weight of 199.70
  • Chemical formula: C6H14ClNO2S
  • Melting/freezing point: 137-138°C
  • Soluble in water and PBS
  • Stable under recommended storage conditions
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.