Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-Methionine methylsulfonium chloride | 3493-12-7 | 100 G
SDP

Catalog No. 5000417996
Encompass_Preferred

DL-methionine methylsulfonium chloride is a naturally occurring methionine derivative that protects gastric mucosal from ethanol-induced damage. It is intended for research use only and not for patient administration. This compound has a molecular weight of 199.70, a chemical formula of C6H14ClNO2S, and appears as a white to off-white solid. Its initial source is animal milk.

  • Naturally occurring methionine derivative
  • Protects gastric mucosal from ethanol-induced damage
  • For research use only
  • Molecular weight: 199.70
  • Chemical formula: C6H14ClNO2S
  • Appearance: White to off-white solid
  • Source: Animals (milk)

Catalog No. 50-004-17996 Supplier Medchemexpress LLC Supplier No. HYN6655100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-methionine methylsulfonium chloride is a naturally occurring methionine derivative that protects gastric mucosal from ethanol-induced damage. It is intended for research use only and not for patient administration. This compound has a molecular weight of 199.70, a chemical formula of C6H14ClNO2S, and appears as a white to off-white solid. Its initial source is animal milk.

  • Naturally occurring methionine derivative
  • Protects gastric mucosal from ethanol-induced damage
  • For research use only
  • Molecular weight: 199.70
  • Chemical formula: C6H14ClNO2S
  • Appearance: White to off-white solid
  • Source: Animals (milk)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.