Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-Methionine methylsulfonium chloride | 3493-12-7 | MFCD00031700 | 500 G
SDP

Catalog No. 5000420099
Encompass_Preferred

DL-methionine methylsulfonium chloride is a naturally occurring methionine derivative. It protects gastric mucosal from ethanol-induced damage.

  • Naturally occurring methionine derivative
  • Protects gastric mucosal from ethanol-induced damage
  • For research use only
  • Purity: 98.0%
  • Appearance: White to off-white solid
  • Soluble in water

Catalog No. 50-004-20099 Supplier Medchemexpress LLC Supplier No. HYN6655500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-methionine methylsulfonium chloride is a naturally occurring methionine derivative. It protects gastric mucosal from ethanol-induced damage.

  • Naturally occurring methionine derivative
  • Protects gastric mucosal from ethanol-induced damage
  • For research use only
  • Purity: 98.0%
  • Appearance: White to off-white solid
  • Soluble in water

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.