Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Methionine Methylsulfonium Chloride 99.0+%, TCI America™
SDP

Catalog No. M064425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M064425G 25 g
M0644500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M064425G Supplier TCI America Supplier No. M064425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3493-12-7
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00031700
InChI Key MYGVPKMVGSXPCQ-UHFFFAOYNA-N
Synonym (3-Amino-3-carboxypropyl)dimethylsulfonium Chloride, S-Methylmethioninesulfonium Chloride, Vitamin U Chloride
PubChem CID 14220
IUPAC Name (3-amino-3-carboxypropyl)-dimethylsulfanium;chloride
SMILES C[S+](C)CCC(C(=O)O)N.[Cl-]

Specifications

Color White
Quantity 25 g
Formula Weight 199.69
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Methionine Methylsulfonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.