Learn More
Medchemexpress LLC DL-Mevalonolactone 10mM 1mL | 674-26-0 | 10MM 1ML
SDP
Catalog No. 5000244505
Encompass_Preferred
DL-Mevalonolactone (( )-Mevalonolactone Mevalolactone) is the -lactone form of mevalonic acid a precursor in the mevalonate pathway DL-Mevalonolactone is orally active against HMGCR mutation and statin caused myopathy[3 DL-Mevalonolactone induces inflammation and oxidative stress response with decreased mitochondrial membrane potential (MMP) and induces mitochondrial swelling[2 [4
Catalog No. 50-002-44505 Supplier Medchemexpress LLC Supplier No. HY10785510MM 1ML
DL-Mevalonolactone (( )-Mevalonolactone Mevalolactone) is the -lactone form of mevalonic acid a precursor in the mevalonate pathway DL-Mevalonolactone is orally active against HMGCR mutation and statin caused myopathy[3 DL-Mevalonolactone induces inflammation and oxidative stress response with decreased mitochondrial membrane potential (MMP) and induces mitochondrial swelling[2 [4