Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dl-mevalonolactone-4,4,5,5,6,6,6-d7 | 347840-19-1 | MFCD01074169 | 99.3% | 137.18 | C6H3D7O3 | 10 MG
SDP

Catalog No. 5000240824
Encompass_Preferred

DL-Mevalonolactone-d7 is the deuterium-labeled analogue of DL-mevalonolactone (the δ-lactone form of mevalonic acid), provided as a stable-isotope research reagent for metabolic and analytical studies. It is intended for laboratory research use only.

  • Deuterium-labeled analogue (d7) of DL-mevalonolactone.
  • Chemical formula C6H3D7O3 and molecular weight 137.18.
  • High measured purity of 99.3%.
  • Supplied in small-scale research pack sizes (10 MG available).
  • Intended for research use only, not for human or diagnostic use.
  • Stable isotope label suitable for tracer and mass spectrometry studies.

Catalog No. 50-002-40824 Supplier Medchemexpress LLC Supplier No. HY107855S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Mevalonolactone-d7 is the deuterium-labeled analogue of DL-mevalonolactone (the δ-lactone form of mevalonic acid), provided as a stable-isotope research reagent for metabolic and analytical studies. It is intended for laboratory research use only.

  • Deuterium-labeled analogue (d7) of DL-mevalonolactone.
  • Chemical formula C6H3D7O3 and molecular weight 137.18.
  • High measured purity of 99.3%.
  • Supplied in small-scale research pack sizes (10 MG available).
  • Intended for research use only, not for human or diagnostic use.
  • Stable isotope label suitable for tracer and mass spectrometry studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.