Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-mevalonolactone-4,4,5,5,6,6,6-d7 | 347840-19-1 | MFCD01074169 | 99.3% | 137.18 | C6H3D7O3 | 5 MG
SDP

Catalog No. 5000242808
Encompass_Preferred

DL-Mevalonolactone-d7 is the deuterium-labeled form of DL-mevalonolactone, the δ-lactone of mevalonic acid. It is designed for use as a stable isotope reagent or internal standard in metabolic profiling and mass spectrometry applications; intended for research use only.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High purity (99.27%).
  • Molecular weight 137.18 g/mol.
  • Soluble in DMSO (100 mg/mL) and water (50 mg/mL); ultrasonic recommended.
  • Supplied as a 5 mg liquid for easy aliquoting.

Catalog No. 50-002-42808 Supplier Medchemexpress LLC Supplier No. HY107855S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Mevalonolactone-d7 is the deuterium-labeled form of DL-mevalonolactone, the δ-lactone of mevalonic acid. It is designed for use as a stable isotope reagent or internal standard in metabolic profiling and mass spectrometry applications; intended for research use only.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High purity (99.27%).
  • Molecular weight 137.18 g/mol.
  • Soluble in DMSO (100 mg/mL) and water (50 mg/mL); ultrasonic recommended.
  • Supplied as a 5 mg liquid for easy aliquoting.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.