Learn More
Medchemexpress LLC DL-mevalonolactone | 674-26-0 | MFCD00006648 | 99.2% | 130.14 g·mol⁻1 | C6H10O3 | 100 MG
SDP
Catalog No. 5000242437
Encompass_Preferred
DL-mevalonolactone is the δ-lactone form of mevalonic acid used as a biochemical research reagent. It is applied in studies of the mevalonate pathway and mitochondrial function and is reported to affect mitochondrial membrane potential and induce oxidative stress in experimental systems.
  • Delta-lactone form of mevalonic acid
  • Used to study the mevalonate pathway and HMGCR-related processes
  • Reported to decrease mitochondrial membrane potential and induce oxidative stress
  • Molecular formula C6H10O3; molecular weight 130.14 g·mol⁻1
  • Reported purity 99.2%
  • Available as solid (multiple weights) and as 10 mM solution in DMSO
Catalog No. 50-002-42437 Supplier Medchemexpress LLC Supplier No. HY107855100MG
DL-mevalonolactone is the δ-lactone form of mevalonic acid used as a biochemical research reagent. It is applied in studies of the mevalonate pathway and mitochondrial function and is reported to affect mitochondrial membrane potential and induce oxidative stress in experimental systems.
  • Delta-lactone form of mevalonic acid
  • Used to study the mevalonate pathway and HMGCR-related processes
  • Reported to decrease mitochondrial membrane potential and induce oxidative stress
  • Molecular formula C6H10O3; molecular weight 130.14 g·mol⁻1
  • Reported purity 99.2%
  • Available as solid (multiple weights) and as 10 mM solution in DMSO