Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-mevalonolactone | 674-26-0 | MFCD00006648 | 99.2% | 130.14 g/mol | C6H10O3 | 250 MG
SDP

Supplier:  Medchemexpress LLC HY107855250MG

Encompass_Preferred

DL-Mevalonolactone is the δ-lactone form of mevalonic acid and an endogenous metabolite in the mevalonate pathway. Supplied as a research-grade chemical, it is used in biochemical and cell-based studies to investigate cholesterol biosynthesis, mitochondrial responses, and related pathway biology.

  • High reported purity approximately 99.2%.
  • Molecular weight 130.14 g/mol.
  • Chemical formula C6H10O3.
  • Available in multiple pack sizes, including a 250 mg solid format.
  • Suitable for biochemical assays, pathway studies, and mitochondrial function experiments.

Catalog No. 50-002-42368


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.