Learn More
Medchemexpress LLC DL-mevalonolactone | 674-26-0 | MFCD00006648 | 99.2% | 130.14 g/mol | C6H10O3 | 500 MG
SDP
Catalog No. 5000240823
Encompass_Preferred
DL-Mevalonolactone is the δ-lactone form of mevalonic acid, an endogenous metabolite and precursor in the mevalonate pathway used in biochemical and pharmacological research. It is employed in studies of cholesterol biosynthesis, HMGCR-related mechanisms, statin-induced myopathy, and assays of mitochondrial function and oxidative stress.
  • Endogenous metabolite involved in the mevalonate biosynthesis pathway.
  • Used in studies of HMGCR function and statin-related myopathy.
  • Applicable to mitochondrial membrane potential and oxidative stress assays.
  • Provided as a high-purity reagent suitable for analytical and biological research.
  • Stable when stored according to supplier recommendations for pure compounds.
Catalog No. 50-002-40823 Supplier Medchemexpress LLC Supplier No. HY107855500MG
DL-Mevalonolactone is the δ-lactone form of mevalonic acid, an endogenous metabolite and precursor in the mevalonate pathway used in biochemical and pharmacological research. It is employed in studies of cholesterol biosynthesis, HMGCR-related mechanisms, statin-induced myopathy, and assays of mitochondrial function and oxidative stress.
  • Endogenous metabolite involved in the mevalonate biosynthesis pathway.
  • Used in studies of HMGCR function and statin-related myopathy.
  • Applicable to mitochondrial membrane potential and oxidative stress assays.
  • Provided as a high-purity reagent suitable for analytical and biological research.
  • Stable when stored according to supplier recommendations for pure compounds.