Learn More
Medchemexpress LLC Dl-norepinephrine-d6 hydrochloride | 1219803-04-9 | 1 MG
SDP
Catalog No. 5000417589
Encompass_Preferred
DL-Norepinephrine-d6 hydrochloride is the deuterium labeled DL-Norepinephrine hydrochloride. It is an external racemic catecholamine neurotransmitter, representing an equal mixture of L- and D-stereoisomers. It can be used as a tracer for Parkinson's disease research when labeled with tritium.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-17589 Supplier Medchemexpress LLC Supplier No. HYN7142S1MG
DL-Norepinephrine-d6 hydrochloride is the deuterium labeled DL-Norepinephrine hydrochloride. It is an external racemic catecholamine neurotransmitter, representing an equal mixture of L- and D-stereoisomers. It can be used as a tracer for Parkinson's disease research when labeled with tritium.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS