Learn More
Selleck Chemical LLC DL-Norepinephrine hydrochloride 5mg 55-27-6 noradrenaline hydrochloride
SDP
Supplier:  Selleck Chemical LLC S57505MG
Encompass_Preferred
Norepinephrine (noradrenaline) is the neurotransmitter at most sympathetic neuroeffector junctions and has pharmacologic effects on both ?1 and ?1 adrenoceptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8407