Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-Norepinephrine hydrochloride

Catalog No. A72561G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A72561G 1 g
A72565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A72561G Supplier Sigma Aldrich Supplier No. A72561G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dl-norepinephrine hydrochloride is an adrenergic drug.dl-norepinephrine hydrochloride is suitable adrenergic drug used in the following studies:
  • To investigate the effects of adrenergic drugs on lantern of larval fly
  • To evaluate the ability of α-adrenergic stimulation in the presence of elevated calcium levels to induce delayed after depolarizations and triggered activity in purkinje fibers isolated from cat hearts
  • Simultaneous reduction and surface functionalization of graphene oxide by a one-step poly(norepinephrine) functionalization

Specifications

CAS 55-27-6
Molecular Formula C8H11NO3 -+ HCl
Linear Formula (HO)2C6H3CH(OH)CH2NH2 -+ HCl
MDL Number MFCD00012880
Quantity 1 g
Synonym (+/-)-1-(3,4-Dihydroxyphenyl)-2-aminoethanol hydrochloride; (+/-)-4-(2-Amino-1-hydroxyethyl)-1,2-benzenediol hydrochloride; DL-Arterenol hydrochloride; DL-Noradrenaline hydrochloride
Molecular Weight (g/mol) 205.64
Percent Purity ≥97% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.