Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Norleucine, 98%, Thermo Scientific Chemicals

Catalog No. AAA1079122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1079122 100 g
AAA1079114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1079122 Supplier Thermo Scientific Chemicals Supplier No. A1079122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-06-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064422
InChI Key LRQKBLKVPFOOQJ-UHFFFAOYNA-N
Synonym dl-norleucine, norleucine, h-dl-nle-oh, norleucine, dl, 2-aminohexanoic acid, dl, dl-2-aminohexanoic acid, dl-.alpha.-aminocaproic acid, norleucine, l, hexanoic acid, s, 2-amino-hexanoic acid
PubChem CID 9475
ChEBI CHEBI:36405
IUPAC Name 2-aminohexanoic acid
SMILES CCCCC(N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721748
Merck Index 14,6706
Quantity 100 g
Odor Odorless
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material DL-Norleucine
EINECSNumber 210-462-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.