Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Norleucine 98.0+%, TCI America™
SDP

Catalog No. N030225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N030225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N030225G Supplier TCI America Supplier No. N030225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-06-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064422
InChI Key LRQKBLKVPFOOQJ-UHFFFAOYNA-N
Synonym dl-norleucine, norleucine, h-dl-nle-oh, norleucine, dl, 2-aminohexanoic acid, dl, dl-2-aminohexanoic acid, dl-.alpha.-aminocaproic acid, norleucine, l, hexanoic acid, s, 2-amino-hexanoic acid
PubChem CID 9475
ChEBI CHEBI:36405
IUPAC Name 2-aminohexanoic acid
SMILES CCCCC(N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Norleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.