Learn More
Apexbio Technology LLC DL-NORMETANEPHRINE-D3-HCL-10MG
SDP
Catalog No. 5000570695
Encompass_Preferred
DL-Normetanephrine-d3 HCl (CAS No 1085333-97-6) is a deuterated form of normetanephrine commonly used in biomedical research for biochemical and pharmacological studies Originating as a stable isotope-labeled analog it is particularly valuable in metabolic and catecholamine pathway investigations This compound acts by influencing neurotransmitter pathways often examined in various receptor binding studies Its in vitro activity is observed in the nanomolar to low micromolar range DL-Normetanephrine-d3 HCl is widely applied in cell models animal studies and drug screening to assess metabolic processes and drug interactions Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70695 Supplier Apexbio Technology LLC Supplier No. SL100510
DL-Normetanephrine-d3 HCl (CAS No 1085333-97-6) is a deuterated form of normetanephrine commonly used in biomedical research for biochemical and pharmacological studies Originating as a stable isotope-labeled analog it is particularly valuable in metabolic and catecholamine pathway investigations This compound acts by influencing neurotransmitter pathways often examined in various receptor binding studies Its in vitro activity is observed in the nanomolar to low micromolar range DL-Normetanephrine-d3 HCl is widely applied in cell models animal studies and drug screening to assess metabolic processes and drug interactions Experimental concentrations typically depend on the specific research design