Learn More
Medchemexpress LLC DL-Norvaline | 760-78-1 | 98.0% | 117.15 | 500 MG
SDP
Catalog No. 5000453495
Encompass_Preferred
DL-Norvaline, also known as 2-Aminopentanoic acid, is a derivative of L-norvaline. L-norvaline acts as a non-competitive inhibitor of arginase. This product is intended for research use only.
  • Functions as an endogenous metabolite.
  • Available as a white to off-white solid.
  • Detailed storage conditions for both powder and solution forms.
  • Solubility information provided for in vitro and in vivo applications.
Catalog No. 50-004-53495 Supplier Medchemexpress LLC Supplier No. HYW010510500MG
DL-Norvaline, also known as 2-Aminopentanoic acid, is a derivative of L-norvaline. L-norvaline acts as a non-competitive inhibitor of arginase. This product is intended for research use only.
  • Functions as an endogenous metabolite.
  • Available as a white to off-white solid.
  • Detailed storage conditions for both powder and solution forms.
  • Solubility information provided for in vitro and in vivo applications.