Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-O-Methylserine 98.0+%, TCI America™
SDP

Catalog No. M08881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08881G 1 g
M08885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08881G Supplier TCI America Supplier No. M08881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19794-53-7
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00037773,MFCD00145257
InChI Key KNTFCRCCPLEUQZ-VKHMYHEASA-N
PubChem CID 97963
IUPAC Name (2S)-2-amino-3-methoxypropanoic acid
SMILES COC[C@H](N)C(O)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 119.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-O-Methylserine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.