Learn More
Cayman Chemical DL-o-Tyrosine, Purity: ≥95%, CAS: 2370-61-8, 500 mg
SDP
Catalog No. 502948030
Encompass_Preferred
A phenylalanine hydroxylation product; has been used as a marker of irradiation processing in poultry meat
Catalog No. 50-294-8030 Supplier Cayman Chemical Supplier No. 33570500 MG
A phenylalanine hydroxylation product; has been used as a marker of irradiation processing in poultry meat