Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-O-tyrosine | 2370-61-8 | MFCD00021725 | 98.2% | 181.19 g/mol | C9H11NO3 | 1 G
SDP

Catalog No. 5000242365
Encompass_Preferred

DL-O-tyrosine (ortho-tyrosine) is the racemic D,L isomer of tyrosine used as an amino acid derivative in biochemical and analytical research. It is employed as a marker of oxidative protein modification and as a building block in synthetic and metabolic studies. The material is supplied as a characterized solid suitable for laboratory use.

  • High purity of 98.2%.
  • Molecular weight 181.19 g/mol.
  • Chemical formula C9H11NO3.
  • White to pink solid appearance.
  • Used as an oxidative stress marker and synthetic intermediate.
  • Available in small laboratory pack sizes such as 1 G.

Catalog No. 50-002-42365 Supplier Medchemexpress LLC Supplier No. HY1078481G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-O-tyrosine (ortho-tyrosine) is the racemic D,L isomer of tyrosine used as an amino acid derivative in biochemical and analytical research. It is employed as a marker of oxidative protein modification and as a building block in synthetic and metabolic studies. The material is supplied as a characterized solid suitable for laboratory use.

  • High purity of 98.2%.
  • Molecular weight 181.19 g/mol.
  • Chemical formula C9H11NO3.
  • White to pink solid appearance.
  • Used as an oxidative stress marker and synthetic intermediate.
  • Available in small laboratory pack sizes such as 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.