Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-o-Tyrosine 98.0+%, TCI America™
SDP

Catalog No. T07861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07861G Supplier TCI America Supplier No. T07861G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unnatural amino acid derivative suitable for chemical biology research with at least 98.0% purity.

Chemical Identifiers

CAS 2370-61-8
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00021725
InChI Key WRFPVMFCRNYQNR-UHFFFAOYNA-N
Synonym dl-o-tyrosine, 2-amino-3-2-hydroxyphenyl propanoic acid, o-tyrosine, 2-hydroxyphenylalanine, 2-hydroxy-dl-phenylalanine, ortho-tyrosine, 2-tyrosine, o-tyrosine, dl, dl-3-o-hydroxyphenyl alanine, 2-amino-3-2-hydroxyphenyl propionic acid
PubChem CID 91482
IUPAC Name 2-amino-3-(2-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=CC=C1O)C(O)=O

Specifications

Color Red-White
Quantity 1 g
Formula Weight 181.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-o-Tyrosine
RTECSNumber YP2285000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.