Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-p-Hydroxyphenyllactic acid

Catalog No. H3253500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H3253500MG 500 mg
H3253 100 mg
H32531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H3253500MG Supplier Sigma Aldrich Supplier No. H3253500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dl-p-hydroxyphenyllactic acid is a precursor in the synthesis of rosmarinic acid that can be used for pharmaceutical and nutraceutical applications
  • It also exhibits antifungal activity.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • H3253.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 306-23-0
Molecular Formula C9H10O4
Linear Formula C9H10O4
MDL Number MFCD00010514
Quantity 500 mg
Synonym 3-(4-Hydroxyphenyl)-2-hydroxypropanoic acid
Molecular Weight (g/mol) 182.17
Percent Purity ≥97% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.