Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-pantolactone | 79-50-5 | 99.3% | 130.14 g/mol | C6H10O3 | 50 G
SDP

Catalog No. 5000210926
Encompass_Preferred

DL-pantolactone is a racemic γ-lactone used as a research chemical and analytical standard. It hydrolyzes to pantoic acid and is used as a synthetic intermediate and analytical reference in biochemical and organic chemistry applications.

  • High purity of 99.3%.
  • Molecular weight 130.14 g/mol.
  • Chemical formula C6H10O3.
  • Solid powder physical form suitable for cold storage.
  • Available in laboratory pack sizes up to 50 g.
  • Commonly used as an analytical standard and synthetic intermediate.

Catalog No. 50-002-10926 Supplier Medchemexpress LLC Supplier No. HYW05351950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-pantolactone is a racemic γ-lactone used as a research chemical and analytical standard. It hydrolyzes to pantoic acid and is used as a synthetic intermediate and analytical reference in biochemical and organic chemistry applications.

  • High purity of 99.3%.
  • Molecular weight 130.14 g/mol.
  • Chemical formula C6H10O3.
  • Solid powder physical form suitable for cold storage.
  • Available in laboratory pack sizes up to 50 g.
  • Commonly used as an analytical standard and synthetic intermediate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.