Learn More
Medchemexpress LLC DL-Pantothenol | 16485-10-2 | 98.0% | 10 MM 1 ML
SDP
Catalog No. 5000413457
Encompass_Preferred
DL-Panthenol (DL-Pantothenol) is a precursor of pantothenic acid. It readily penetrates the skin, offering deep moisturizing properties, promoting wound healing, and acting as an anti-inflammatory agent. This compound is also utilized in cosmetics as a hair conditioner. DL-Panthenol is characterized by its low acute toxicity, non-sensitizing nature, and absence of significant genotoxicity or reproductive and developmental toxicity, making it suitable for cosmetic research applications.
  • Precursor of pantothenic acid.
  • Easily penetrates the skin.
  • Provides deep moisturizing effects.
  • Promotes wound healing.
  • Possesses anti-inflammatory properties.
  • Functions as a hair conditioner in cosmetics.
  • Exhibits low acute toxicity.
  • Non-sensitizing property.
  • No significant genotoxicity.
  • No significant reproductive and developmental toxicity.
Catalog No. 50-004-13457 Supplier Medchemexpress LLC Supplier No. HYB102410MM/1ML
DL-Panthenol (DL-Pantothenol) is a precursor of pantothenic acid. It readily penetrates the skin, offering deep moisturizing properties, promoting wound healing, and acting as an anti-inflammatory agent. This compound is also utilized in cosmetics as a hair conditioner. DL-Panthenol is characterized by its low acute toxicity, non-sensitizing nature, and absence of significant genotoxicity or reproductive and developmental toxicity, making it suitable for cosmetic research applications.
  • Precursor of pantothenic acid.
  • Easily penetrates the skin.
  • Provides deep moisturizing effects.
  • Promotes wound healing.
  • Possesses anti-inflammatory properties.
  • Functions as a hair conditioner in cosmetics.
  • Exhibits low acute toxicity.
  • Non-sensitizing property.
  • No significant genotoxicity.
  • No significant reproductive and developmental toxicity.