Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Penicillamine ≥99%, Thermo Scientific™

Catalog No. AC129751000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC129751000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC129751000 Supplier Thermo Scientific Chemicals Supplier No. 129751000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52-66-4
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C5H11NO2S
Linear Formula (CH3)2C(SH)CH(NH2)CO2H
Quantity 100g
MDL Number MFCD00004856
Merck Index 15, 7197
Synonym dl-penicillamine, 2-amino-3-mercapto-3-methylbutanoic acid, +--penicillamine, 3,3-dimethyl-dl-cysteine, 3-sulfanylvaline, dl-beta-mercaptovaline, beta-mercaptovaline, d,l-penicillamine, dl-3-mercaptovaline, 2-amino-3-mercapto-3-methylbutyric acid
Specific Rotation -1° to +1° (20°C, 589nm) (c=5, 1M NaOH)
InChI Key VVNCNSJFMMFHPL-UHFFFAOYNA-N
SMILES CC(C)(S)C(N)C(O)=O
IUPAC Name 2-amino-3-methyl-3-sulfanylbutanoic acid
Molecular Weight (g/mol) 149.21
PubChem CID 4727
ChEBI CHEBI:50868
Formula Weight 149.21
Percent Purity ≥96.0%
Chemical Name or Material DL-Penicillamine, 99+%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-147-2
RTECSNumber YV9445000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.