Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Penicillamine 98.0+%, TCI America™
SDP

Catalog No. P00251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P00251G Supplier TCI America Supplier No. P00251G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52-66-4
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00004856
InChI Key VVNCNSJFMMFHPL-UHFFFAOYNA-N
Synonym dl-penicillamine, 2-amino-3-mercapto-3-methylbutanoic acid, +--penicillamine, 3,3-dimethyl-dl-cysteine, 3-sulfanylvaline, dl-beta-mercaptovaline, beta-mercaptovaline, d,l-penicillamine, dl-3-mercaptovaline, 2-amino-3-mercapto-3-methylbutyric acid
PubChem CID 4727
ChEBI CHEBI:50868
IUPAC Name 2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(S)C(N)C(O)=O

Specifications

Melting Point 205°C
Color White
Quantity 1 g
Formula Weight 149.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Penicillamine
RTECSNumber YV9445000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.