Learn More
Cayman Chemical DL-Phenyl-d5-alanine, Purity: ≥99% deuterated forms (d1-d5), CAS: 284664-89-7, 100 mg
SDP
Catalog No. 502911304
Encompass_Preferred
An internal standard for the quantification of DL-phenylalanine by GC- or LC-MS
Catalog No. 50-291-1304 Supplier Cayman Chemical Supplier No. 34845100 MG
An internal standard for the quantification of DL-phenylalanine by GC- or LC-MS