Learn More
AdipoGen DL-Phenylalanine
SDP
Catalog No. 502560206
Encompass_Preferred
Chemical. CAS 150-30-1. Formula C9H11NO2. MW 165.19. Synthetic. Essential alpha-amino acid in humans. Mixture of D-phenylalanine and L-phenylalanine. Precursor for tyrosine, the monoamine neurotransmitters dopamine, norepinephrine noradrenaline, epinephrine adrenaline, the neuromodulator phenethylamine and the skin pigment melanin. Used in the manufacture of food and drink products and sold as a nutritional supplement for its reputed analgesic and antidepressant effects. Building block which may be used by as starting material for synthesis.
Catalog No. 50-256-0206 Supplier AdipoGen Supplier No. CDXP0185G250
Chemical. CAS 150-30-1. Formula C9H11NO2. MW 165.19. Synthetic. Essential alpha-amino acid in humans. Mixture of D-phenylalanine and L-phenylalanine. Precursor for tyrosine, the monoamine neurotransmitters dopamine, norepinephrine noradrenaline, epinephrine adrenaline, the neuromodulator phenethylamine and the skin pigment melanin. Used in the manufacture of food and drink products and sold as a nutritional supplement for its reputed analgesic and antidepressant effects. Building block which may be used by as starting material for synthesis.