Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Phenylephrine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P039725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P039725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P039725G Supplier TCI America Supplier No. P039725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 154-86-9
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.666
MDL Number MFCD00070603
InChI Key OCYSGIYOVXAGKQ-UHFFFAOYSA-N
Synonym 3-[1-Hydroxy-2-(methylamino)ethyl]phenol Hydrochloride, 1-(3-Hydroxyphenyl)-2-(methylamino)ethanol Hydrochloride, 3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol Hydrochloride
PubChem CID 6102
IUPAC Name 3-[1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
SMILES CNCC(C1=CC(=CC=C1)O)O.Cl

Specifications

Melting Point 144°C
Color White
Quantity 25 g
Formula Weight 203.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Phenylephrine Hydrochloride
RTECSNumber DO7600000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.