Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Phenylmercapturic Acid 98.0+%, TCI America™
SDP

Catalog No. P06075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06075G Supplier TCI America Supplier No. P06075G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20640-68-0
Molecular Formula C11H13NO3S
Molecular Weight (g/mol) 239.289
MDL Number MFCD00059335
InChI Key CICOZWHZVMOPJS-UHFFFAOYSA-N
Synonym N-Acetyl-S-phenyl-DL-cysteine, Ac-DL-Cys(Ph)-OH
PubChem CID 95526
IUPAC Name 2-acetamido-3-phenylsulfanylpropanoic acid
SMILES CC(=O)NC(CSC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 239.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Phenylmercapturic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.