Learn More
Alkali Scientific DL PHOSPHINOTHRICIN [PPT], 250 Mg
SDP
Catalog No. 5000660222
Encompass_Preferred
Phosphinothricin (PPT) is an amino acid analog and a high-purity biochemical reagent used to inhibit glutamine synthetase in both plant and microbial systems. It is commonly utilized in genetic research to select for resistant organisms in plant transformation and bacterial studies. PPT functions by preventing the synthesis of glutamine, an essential amino acid, which disrupts metabolic processes in cells. This reagent is available in a 250mg quantity and is used in agricultural research, genetic engineering, and molecular biology to study gene expression, enzymatic inhibition, and to select genetically modified organisms (GMOs). It is also used in herbicide resistance studies.
Catalog No. 50-006-60222 Supplier Alkali Scientific Supplier No. DLP001CP
Phosphinothricin (PPT) is an amino acid analog and a high-purity biochemical reagent used to inhibit glutamine synthetase in both plant and microbial systems. It is commonly utilized in genetic research to select for resistant organisms in plant transformation and bacterial studies. PPT functions by preventing the synthesis of glutamine, an essential amino acid, which disrupts metabolic processes in cells. This reagent is available in a 250mg quantity and is used in agricultural research, genetic engineering, and molecular biology to study gene expression, enzymatic inhibition, and to select genetically modified organisms (GMOs). It is also used in herbicide resistance studies.